siRNA Details
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026933.2 | 10, 11 | 1725 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026933.2 | 10, 11 | 1725 |