siRNA Details
| Gene Symbol: LOC101927865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI925949.1 | - | 3 | 162 | ||
| DB075298.1 | - | 5 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927865| Gene Symbol: LOC101927865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI925949.1 | - | 3 | 162 | ||
| DB075298.1 | - | 5 | 371 | |||