siRNA Details
Gene Symbol: PRR23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013650.2 | NP_001013672.1 | 1 | 953 | ||
GenBank mRNA | AK127998.1 | - | 1 | 953 | ||
BC137145.1 | - | 1 | 941 | 9021142 | ||
BC137146.1 | - | 1 | 939 | 9021143 |
Gene Symbol: PRR23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134659.1 | NP_001128131.1 | 1 | 691 |
Gene Symbol: PRR23C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134657.1 | NP_001128129.1 | 1 | 960 | ||
GenBank mRNA | AK128089.1 | - | 1 | 957 |
