siRNA Details
| Gene Symbol: PACRG-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122120.1 | 3 | 246 | |||
| GenBank mRNA | AK311212.1 | - | 3 | 246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PACRG-AS3| Gene Symbol: PACRG-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122120.1 | 3 | 246 | |||
| GenBank mRNA | AK311212.1 | - | 3 | 246 | ||