siRNA Details
| Gene Symbol: XCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024644.1 | NP_001019815.1 | 2 | 415 | ||
| NM_005283.2 | NP_005274.1 | 3 | 537 | |||
| XM_017006196.1 | XP_016861685.1 | 4 | 1555 | |||
| XM_017006197.1 | XP_016861686.1 | 6 | 1807 | |||
| XM_017006198.1 | XP_016861687.1 | 5 | 1620 | |||
| XM_017006199.1 | XP_016861688.1 | 4 | 617 | |||
| XM_017006200.1 | XP_016861689.1 | 3 | 1475 | |||
| XM_017006201.1 | XP_016861690.1 | 2 | 1324 | |||
| GenBank mRNA | AK314277.1 | - | 2 | 668 | ||
| BC069075.1 | - | 1 | 313 | 7216889 | ||
| BC069589.1 | - | 1 | 313 | 7262280 | ||
| BC095533.1 | - | 1 | 364 | 6971949 | ||

XCR1
Genomic Map