siRNA Details
| Gene Symbol: CEACAM18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278392.1 | NP_001265321.1 | 2 | 316 | ||
| XM_011527258.2 | XP_011525560.1 | 2 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM18| Gene Symbol: CEACAM18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278392.1 | NP_001265321.1 | 2 | 316 | ||
| XM_011527258.2 | XP_011525560.1 | 2 | 416 | |||