siRNA Details
| Gene Symbol: CXCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142797.1 | NP_001136269.1 | 2 | 1840 | ||
| NM_001504.1 | NP_001495.1 | 2 | 1596 | |||
| XM_005262256.3 | XP_005262313.1 | 2 | 1697 | |||
| XM_005262257.3 | XP_005262314.1 | 3 | 1360 | |||
| XM_017029435.1 | XP_016884924.1 | 2 | 1778 | |||
| XM_017029436.1 | XP_016884925.1 | 3 | 1257 | |||
| GenBank mRNA | AF469635.1 | - | 2 | 1833 | ||
| BC034403.1 | - | 2 | 1566 | 5176136 | ||
| X95876.1 | - | 2 | 1596 | |||

CXCR3
Genomic Map