siRNA Details
| Gene Symbol: LOC107983997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011527096.2 | XP_011525398.1 | 4 | 608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107983997| Gene Symbol: LOC107983997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011527096.2 | XP_011525398.1 | 4 | 608 | ||