siRNA Details
| Gene Symbol: LSP1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033961.1 | 1 | 232 | |||
| GenBank mRNA | BC121073.2 | - | 1 | 236 | 40119616 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSP1P3| Gene Symbol: LSP1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033961.1 | 1 | 232 | |||
| GenBank mRNA | BC121073.2 | - | 1 | 236 | 40119616 | |