siRNA Details
Gene Symbol: GPR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005290.3 | NP_005281.1 | 1 | 581 | ||
GenBank mRNA | AK313375.1 | - | 1 | 581 | ||
BC069437.1 | - | 1 | 446 | 7262234 | ||
BC101779.1 | - | 1 | 472 | 8069285 | ||
BC101781.1 | - | 1 | 469 | 8069287 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
