siRNA Details
| Gene Symbol: SKP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514081.2 | XP_011512383.1 | 1 | 40 | ||
| XM_011514082.2 | XP_011512384.1 | 1 | 40 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKP2| Gene Symbol: SKP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514081.2 | XP_011512383.1 | 1 | 40 | ||
| XM_011514082.2 | XP_011512384.1 | 1 | 40 | |||