siRNA Details
| Gene Symbol: RFPL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145014.1 | NP_001138486.1 | 3 | 771 | ||
| XM_011526915.2 | XP_011525217.1 | 4 | 2059 | |||
| Gene Symbol: RFPL4AL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277397.1 | NP_001264326.1 | 2 | 727 | ||
| XM_011527259.1 | XP_011525561.1 | 3 | 794 | |||

RFPL4A
Genomic Map