siRNA Details
Gene Symbol: GPR21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005294.2 | NP_005285.1 | 2 | 1173 | ||
XM_005251933.3 | XP_005251990.1 | 2 | 7380 | |||
GenBank mRNA | AK299686.1 | - | 2 | 1166 | ||
AK313539.1 | - | 2 | 1147 | |||
BC066885.1 | - | 1 | 677 | 6971731 | ||
BC066886.1 | - | 1 | 677 | 6971734 | ||
BC067532.1 | - | 1 | 677 | 6971732 | ||
BC067533.1 | - | 1 | 677 | 6971733 | ||
BC067534.1 | - | 1 | 677 | 6971735 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
