siRNA Details
| Gene Symbol: LOC102724344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135182.1 | 1 | 24 | |||
| NR_135183.1 | 1 | 24 | ||||
| GenBank mRNA | DB046859.1 | - | 1 | 24 | ||
| DB057778.1 | - | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724344| Gene Symbol: LOC102724344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135182.1 | 1 | 24 | |||
| NR_135183.1 | 1 | 24 | ||||
| GenBank mRNA | DB046859.1 | - | 1 | 24 | ||
| DB057778.1 | - | 1 | 23 | |||