siRNA Details
| Gene Symbol: LOC441178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117092.1 | 4 | 819 | |||
| GenBank mRNA | AK056600.1 | - | 4 | 819 | ||
| AL832737.1 | - | 6 | 2151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441178| Gene Symbol: LOC441178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117092.1 | 4 | 819 | |||
| GenBank mRNA | AK056600.1 | - | 4 | 819 | ||
| AL832737.1 | - | 6 | 2151 | |||