siRNA Details
| Gene Symbol: ZNF418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317028.1 | NP_001303957.1 | 2 | 71 | ||
| NM_001317029.1 | NP_001303958.1 | 2 | 71 | |||
| XM_017026310.1 | XP_016881799.1 | 2 | 138 | |||
| XM_017026311.1 | XP_016881800.1 | 2 | 138 | |||
| XM_017026313.1 | XP_016881802.1 | 2 | 138 | |||
| XM_017026314.1 | XP_016881803.1 | 2 | 137 | |||
| GenBank mRNA | DA113968.1 | - | 2 | 57 | ||
| DA961678.1 | - | 2 | 77 | |||

ZNF418
Genomic Map