siRNA Details
| Gene Symbol: GPR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018971.1 | NP_061844.1 | 1 | 828 | ||
| GenBank mRNA | AB040799.1 | - | 1 | 828 | ||
| HQ709187.1 | - | 1 | 828 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR27| Gene Symbol: GPR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018971.1 | NP_061844.1 | 1 | 828 | ||
| GenBank mRNA | AB040799.1 | - | 1 | 828 | ||
| HQ709187.1 | - | 1 | 828 | |||
Set Membership: |
|