siRNA Details
| Gene Symbol: C16orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164579.1 | NP_001158051.1 | 2 | 301 | ||
| GenBank mRNA | BC027604.1 | - | 3 | 331 | ||
| BC053950.1 | - | 3 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf52| Gene Symbol: C16orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164579.1 | NP_001158051.1 | 2 | 301 | ||
| GenBank mRNA | BC027604.1 | - | 3 | 331 | ||
| BC053950.1 | - | 3 | 318 | |||