siRNA Details
| Gene Symbol: TMEM229A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136002.1 | NP_001129474.1 | 1 | 1517 | ||
| GenBank mRNA | BC157828.1 | - | 1 | 1068 | 9057047 | |
| BC171809.1 | - | 1 | 1068 | 9054463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM229A| Gene Symbol: TMEM229A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136002.1 | NP_001129474.1 | 1 | 1517 | ||
| GenBank mRNA | BC157828.1 | - | 1 | 1068 | 9057047 | |
| BC171809.1 | - | 1 | 1068 | 9054463 | ||