siRNA Details
| Gene Symbol: LOC102724699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125409.1 | 2 | 388 | |||
| GenBank mRNA | AK094454.1 | - | 2 | 388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724699| Gene Symbol: LOC102724699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125409.1 | 2 | 388 | |||
| GenBank mRNA | AK094454.1 | - | 2 | 388 | ||