siRNA Details
| Gene Symbol: GPR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195381.1 | NP_001182310.1 | 6 | 1473 | ||
| NM_001195382.1 | NP_001182311.1 | 6 | 1359 | |||
| NM_005301.3 | NP_005292.2 | 2 | 836 | |||
| GenBank mRNA | AF089087.1 | - | 1 | 1083 | ||
| AK131540.1 | - | 1 | 3019 | |||
| AK172786.1 | - | 6 | 1450 | |||
| AK290256.1 | - | 6 | 1322 | |||
| AK314002.1 | - | 2 | 836 | |||
| BC095500.1 | - | 1 | 505 | |||
| BC117453.1 | - | 1 | 880 | 40126004 | ||
| BC117455.1 | - | 1 | 878 | 40126006 | ||
| CR541765.1 | - | 1 | 508 | |||

GPR35
Genomic Map