siRNA Details
| Gene Symbol: LOC100507073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA610966.1 | - | 1 | 61 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507073| Gene Symbol: LOC100507073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA610966.1 | - | 1 | 61 | ||