siRNA Details
| Gene Symbol: LOC101929241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110166.1 | 1, 2 | 106 | |||
| GenBank mRNA | DB124165.1 | - | 1, 2 | 106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929241| Gene Symbol: LOC101929241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110166.1 | 1, 2 | 106 | |||
| GenBank mRNA | DB124165.1 | - | 1, 2 | 106 | ||