siRNA Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF469199.1 | - | 2 | 178 | ||
| AF469200.1 | - | 2 | 178 | |||
| AF469204.1 | - | 1 | 26 | |||
| BC065701.1 | - | 3 | 380 | |||
| BC111049.1 | - | 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIO3OS| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF469199.1 | - | 2 | 178 | ||
| AF469200.1 | - | 2 | 178 | |||
| AF469204.1 | - | 1 | 26 | |||
| BC065701.1 | - | 3 | 380 | |||
| BC111049.1 | - | 2 | 197 | |||