siRNA Details
| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001508.2 | NP_001499.1 | 2 | 2695 | ||
| GenBank mRNA | AK122643.1 | - | 2 | 2695 | ||
| AK123759.1 | - | 3 | 1929 | |||
| DA653793.1 | - | 1 | 427 | |||
More Information
Set Membership: |
|

GPR39
Genomic Map