siRNA Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005304.3 | NP_005295.1 | 2 | 139 | ||
| XM_011526858.2 | XP_011525160.1 | 2 | 495 | |||
| GenBank mRNA | AK290710.1 | - | 2 | 139 | ||
| BC035657.1 | - | 2 | 125 | 5588752 | ||
| BC113695.1 | - | 1 | 134 | 8322747 | ||
| BC148269.1 | - | 1 | 86 | 8860212 | ||
| EU432115.1 | - | 1 | 64 | |||

FFAR3
Genomic Map