siRNA Details
| Gene Symbol: LOC730338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134575.1 | 2, 3 | 276 | |||
| GenBank mRNA | AK125311.1 | - | 2, 3 | 276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC730338| Gene Symbol: LOC730338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134575.1 | 2, 3 | 276 | |||
| GenBank mRNA | AK125311.1 | - | 2, 3 | 276 | ||