siRNA Details
| Gene Symbol: LINC01556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103538.1 | 1 | 677 | |||
| GenBank mRNA | BX118328.1 | - | 1 | 677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01556| Gene Symbol: LINC01556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103538.1 | 1 | 677 | |||
| GenBank mRNA | BX118328.1 | - | 1 | 677 | ||