siRNA Details
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000740.2 | NP_000731.1 | 5 | 2338 | ||
XM_005273032.3 | XP_005273089.1 | 6 | 2643 | |||
XM_011544041.2 | XP_011542343.1 | 7 | 2742 | |||
XM_011544043.2 | XP_011542345.1 | 9 | 3075 | |||
XM_011544044.2 | XP_011542346.1 | 8 | 2846 | |||
XM_011544045.2 | XP_011542347.1 | 8 | 2971 | |||
XM_011544046.2 | XP_011542348.1 | 6 | 2679 | |||
XM_011544047.2 | XP_011542349.1 | 6 | 2615 | |||
XM_017000152.1 | XP_016855641.1 | 8 | 5668 | |||
XM_017000153.1 | XP_016855642.1 | 7 | 2746 | |||
XM_017000154.1 | XP_016855643.1 | 6 | 2440 | |||
XM_017000155.1 | XP_016855644.1 | 7 | 2783 | |||
XM_017000156.1 | XP_016855645.1 | 7 | 2747 | |||
XM_017000157.1 | XP_016855646.1 | 5 | 2348 | |||
XM_017000158.1 | XP_016855647.1 | 5 | 2580 | |||
XM_017000159.1 | XP_016855648.1 | 5 | 2552 | |||
XM_017000160.1 | XP_016855649.1 | 4 | 3402 | |||
XM_017000161.1 | XP_016855650.1 | 6 | 2684 | |||
XM_017000162.1 | XP_016855651.1 | 5 | 2516 | |||
XM_017000163.1 | XP_016855652.1 | 4 | 6378 | |||
GenBank mRNA | AF385589.1 | - | 1 | 1559 | ||
AF498917.1 | - | 1 | 1559 | |||
AK074575.1 | - | 1 | 1716 | |||
BC096844.1 | - | 1 | 1560 | |||
BC121026.2 | - | 2 | 1702 | 40117429 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_033269, BC129893, AK080950, BC129892.
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
