siRNA Details
| Gene Symbol: TGIF2LY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139214.2 | NP_631960.1 | 2 | 357 | ||
| GenBank mRNA | AF332223.1 | - | 2 | 357 | ||
| AK292609.1 | - | 2 | 401 | |||
| BC128604.1 | - | 1 | 327 | 40003247 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TGIF2LY| Gene Symbol: TGIF2LY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139214.2 | NP_631960.1 | 2 | 357 | ||
| GenBank mRNA | AF332223.1 | - | 2 | 357 | ||
| AK292609.1 | - | 2 | 401 | |||
| BC128604.1 | - | 1 | 327 | 40003247 | ||