siRNA Details
| Gene Symbol: TBC1D3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025494.1 | XP_016880983.1 | 3, 4 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D3J| Gene Symbol: TBC1D3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025494.1 | XP_016880983.1 | 3, 4 | 196 | ||