siRNA Details
| Gene Symbol: LOC107984139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027560.1 | XP_016883049.1 | 2 | 146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107984139| Gene Symbol: LOC107984139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027560.1 | XP_016883049.1 | 2 | 146 | ||