siRNA Details
| Gene Symbol: OR2T34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001821.1 | NP_001001821.1 | 1 | 281 | ||
| GenBank mRNA | BC136924.1 | - | 1 | 303 | 9020916 | |
| BC136926.1 | - | 1 | 303 | 9020918 | ||
| Gene Symbol: OR2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005495.1 | NP_001005495.1 | 1 | 281 | ||
| GenBank mRNA | BC136915.1 | - | 1 | 306 | 9020906 | |
| BC136916.1 | - | 1 | 306 | 9020907 | ||

OR2T34
Genomic Map