siRNA Details
| Gene Symbol: CSN1S2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033311.1 | 5, 6 | 189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSN1S2BP| Gene Symbol: CSN1S2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033311.1 | 5, 6 | 189 | |||