siRNA Details
| Gene Symbol: MS4A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310141.1 | NP_001297070.1 | 2 | 969 | ||
| XM_011545420.2 | XP_011543722.1 | 2 | 2067 | |||
| XM_011545421.2 | XP_011543723.1 | 2 | 2066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MS4A18| Gene Symbol: MS4A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310141.1 | NP_001297070.1 | 2 | 969 | ||
| XM_011545420.2 | XP_011543722.1 | 2 | 2067 | |||
| XM_011545421.2 | XP_011543723.1 | 2 | 2066 | |||