siRNA Details
| Gene Symbol: DCDC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287444.1 | NP_001274373.1 | 9 | 1130 | ||
| XM_006711894.3 | XP_006711957.1 | 9 | 1124 | |||
| XM_017004836.1 | XP_016860325.1 | 9 | 1124 | |||
| XM_017004837.1 | XP_016860326.1 | 9 | 1124 | |||
| XM_017004838.1 | XP_016860327.1 | 8 | 995 | |||

DCDC2C
Genomic Map