siRNA Details
Gene Symbol: LINC00301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026946.1 | 7 | 687 | |||
GenBank mRNA | AK058123.1 | - | 7 | 687 | ||
BC029583.1 | - | 6 | 570 | |||
BX103095.1 | - | 6 | 573 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026946.1 | 7 | 687 | |||
GenBank mRNA | AK058123.1 | - | 7 | 687 | ||
BC029583.1 | - | 6 | 570 | |||
BX103095.1 | - | 6 | 573 |