siRNA Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297746.1 | NP_001284675.1 | 3 | 408 | ||
| NM_016076.4 | NP_057160.2 | 4 | 507 | |||
| NR_123735.1 | 4 | 502 | ||||
| NR_123736.1 | 3 | 413 | ||||
| XM_011544203.2 | XP_011542505.1 | 5 | 592 | |||
| XM_011544205.2 | XP_011542507.1 | 4 | 1158 | |||
| XM_011544206.2 | XP_011542508.1 | 4 | 231 | |||
| XM_011544207.2 | XP_011542509.1 | 3 | 1598 | |||
| GenBank mRNA | AF151904.1 | - | 4 | 299 | ||
| AF229834.1 | - | 3 | 149 | |||
| AK314664.1 | - | 4 | 477 | |||
| BC004485.2 | - | 4 | 454 | 3831754 | ||
| BI459676.1 | - | 4 | 510 | |||
| CB854052.1 | - | 3 | 165 | |||
| CN303012.1 | - | 4 | 413 | |||

DESI2
Genomic Map