siRNA Details
| Gene Symbol: ZNF735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159524.1 | NP_001152996.1 | 4 | 1011 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF735| Gene Symbol: ZNF735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159524.1 | NP_001152996.1 | 4 | 1011 | ||