siRNA Details
| Gene Symbol: ZNF860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137674.2 | NP_001131146.2 | 2 | 1521 | ||
| XM_017006299.1 | XP_016861788.1 | 2 | 2604 | |||
| GenBank mRNA | AK294003.1 | - | 2 | 1521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF860| Gene Symbol: ZNF860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137674.2 | NP_001131146.2 | 2 | 1521 | ||
| XM_017006299.1 | XP_016861788.1 | 2 | 2604 | |||
| GenBank mRNA | AK294003.1 | - | 2 | 1521 | ||