siRNA Details
| Gene Symbol: LOC101927016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011535339.1 | XP_011533641.1 | 1 | 1514 | ||
| Gene Symbol: LINC01198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC046635.1 | - | 1 | 602 | ||

LOC101927016
Genomic Map