siRNA Details
| Gene Symbol: IGSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529472.1 | XP_011527774.1 | 2 | 1351 | ||
| XM_011529473.1 | XP_011527775.1 | 2 | 1351 | |||
| XM_017028279.1 | XP_016883768.1 | 2 | 1351 | |||
| XM_017028280.1 | XP_016883769.1 | 2 | 1351 | |||
| Gene Symbol: LOC732226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX396897.2 | - | 1 | 113 | ||

IGSF5
Genomic Map