siRNA Details
| Gene Symbol: GRIK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000831.3 | NP_000822.2 | 2 | 376 | ||
| GenBank mRNA | AJ249210.1 | - | 2 | 262 | ||
| AJ299451.1 | - | 2 | 241 | |||
| BI544689.1 | - | 380 | ||||
| DR003075.1 | - | 2 | 560 | |||
| U16127.1 | - | 2 | 259 | |||

GRIK3
Genomic Map