siRNA Details
| Gene Symbol: LINC00917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024406.1 | 7 | 1855 | |||
| GenBank mRNA | AK093244.1 | - | 7 | 1855 | ||
| BC043250.1 | - | 6 | 1912 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00917| Gene Symbol: LINC00917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024406.1 | 7 | 1855 | |||
| GenBank mRNA | AK093244.1 | - | 7 | 1855 | ||
| BC043250.1 | - | 6 | 1912 | |||