siRNA Details
| Gene Symbol: VRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288837.1 | NP_001275766.1 | 1 | 178 | ||
| NM_001288838.1 | NP_001275767.1 | 1 | 178 | |||
| GenBank mRNA | AK292727.1 | - | 1 | 164 | ||
| AK307113.1 | - | 1 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VRK2| Gene Symbol: VRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288837.1 | NP_001275766.1 | 1 | 178 | ||
| NM_001288838.1 | NP_001275767.1 | 1 | 178 | |||
| GenBank mRNA | AK292727.1 | - | 1 | 164 | ||
| AK307113.1 | - | 1 | 177 | |||