siRNA Details
| Gene Symbol: LOC105374250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322103.1 | NP_001309032.1 | 4 | 1017 | ||
| XM_011513348.2 | XP_011511650.1 | 3 | 836 | |||
| GenBank mRNA | BG190345.1 | - | 1 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374250| Gene Symbol: LOC105374250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322103.1 | NP_001309032.1 | 4 | 1017 | ||
| XM_011513348.2 | XP_011511650.1 | 3 | 836 | |||
| GenBank mRNA | BG190345.1 | - | 1 | 265 | ||