siRNA Details
| Gene Symbol: LOC285074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057663.1 | - | 7 | 1912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285074| Gene Symbol: LOC285074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057663.1 | - | 7 | 1912 | ||