siRNA Details
| Gene Symbol: FBXW10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267585.1 | NP_001254514.1 | 14 | 2872 | ||
| NM_001267586.1 | NP_001254515.1 | 13 | 2713 | |||
| GenBank mRNA | AL136822.1 | - | 13 | 2651 | ||
| AY729024.1 | - | 14 | 2640 | |||
| BC028364.2 | - | 12 | 2886 | 4827344 | ||
| Gene Symbol: CDRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282540.1 | NP_001269469.1 | 13 | 2640 | ||
| GenBank mRNA | AF337810.1 | - | 1 | 225 | ||

FBXW10
Genomic Map