siRNA Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104192.2 | - | 1 | 308 | ||
| BC104193.2 | - | 1 | 308 | |||
| BC112986.1 | - | 1 | 308 | |||
| BC112987.1 | - | 1 | 308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTGER4P2-CDK2AP2P2| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104192.2 | - | 1 | 308 | ||
| BC104193.2 | - | 1 | 308 | |||
| BC112986.1 | - | 1 | 308 | |||
| BC112987.1 | - | 1 | 308 | |||