siRNA Details
| Gene Symbol: ZNF596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287255.1 | NP_001274184.1 | 1 | 44 | ||
| NM_001287256.1 | NP_001274185.1 | 1 | 44 | |||
| GenBank mRNA | DB034950.1 | - | 1 | 44 | ||
| DB055646.1 | - | 1 | 13 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF596| Gene Symbol: ZNF596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287255.1 | NP_001274184.1 | 1 | 44 | ||
| NM_001287256.1 | NP_001274185.1 | 1 | 44 | |||
| GenBank mRNA | DB034950.1 | - | 1 | 44 | ||
| DB055646.1 | - | 1 | 13 | |||